Z-Ala-Ala-OH 98%

Supplier: Ambeed
Warning

A455279-1G A455279-25G A455279-5G
76789-412EA 10 USD
76789-412 76789-414 77572-436
Z-Ala-Ala-OH 98%
Z-Ala-Ala-OH

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₅
MDL Number: MFCD00037234
CAS Number: 16012-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR