Tetrahydropyran-2-methanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxymethyltetrahydropyran, 2-(Hydroxymethyl)tetrahydro-2H-pyran

A454940-1G A454940-5G A454940-25G
76789-406EA 19.1 USD
76789-406 76789-408 77601-960
Tetrahydropyran-2-methanol 98%
Tetrahydropyran-2-methanol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00006624
CAS Number: 100-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR