4-Iodo-3-hydroxypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Iodopyridin-3-ol

A454050-100MG A454050-10G A454050-1G A454050-250MG A454050-5G
76789-380EA 19.1 USD
76789-380 76789-382 76789-384 76789-386 76789-388
4-Iodo-3-hydroxypyridine 95%
4-Iodo-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄INO
MW: 221 g/mol
MDL Number: MFCD09030242
CAS Number: 188057-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR