(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone 95%

Supplier: Ambeed
Warning

A453838-100MG A453838-50MG
76789-376EA 249.32 USD
76789-376 76789-378
(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone 95%
(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₉ClFIO₂
MW: 390.576 g/mol
MDL Number: MFCD11520937
CAS Number: 1233025-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR