Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride 97%

Supplier: Ambeed
Warning

Synonyms: Di-μ-hydroxo-bis[(N,N,N'.N'-tetramethylethylenediamine)copper(II)] chloride

A453156-5G A453156-10G A453156-100G A453156-1G A453156-25G
76789-366EA 54.11 USD
76789-366 77572-428 77669-036 77669-038 77669-040
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride 97%
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride

Precaución: for Research Use Only

Formula: C₁₂H₃₄Cl₂Cu₂N₄O₂
MW: 464.42 g/mol
MDL Number: MFCD01321163
CAS Number: 30698-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR