Zinc hydroxide 97%

Supplier: Ambeed
Danger

Synonyms: Zinc Dihydroxide, Zinchydroxide, Zinc (²⁺) ion dioxidanide

A451977-25G A451977-1000G A451977-100G A451977-500G
76789-364EA 18 USD
76789-364 77572-420 77669-028 77669-030
Zinc hydroxide 97%
Zinc hydroxide

Precaución: for Research Use Only

Formula: H₂O₂Zn
MDL Number: MFCD00049620
CAS Number: 20427-58-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR