Trichloromethiazide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
6-Chloro-3-(dichloromethyl)-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulphonamide 1,1-dioxide, 6-Chloro-3-dichloromethyl-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulphonamide 1,1-Dioxide
A451003-1G
A451003-1MG
A451003-250MG
A451003-25G
A451003-5G
76789-354EA
16.98
USD
76789-354
76789-356
76789-358
76789-360
76789-362
Trichloromethiazide 98%
Trichloromethiazide
Precaución: for Research Use Only
Formula:
C₈H₈Cl₃N₃O₄S₂ MW: 380.64 g/mol |
MDL Number:
MFCD00057315 CAS Number: 133-67-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...