2-Chloro-(5-methoxycarbonyl)phenylboronic acid 96%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-5-(methoxycarbonyl)phenylboronic acid, (2-Chloro-5-(methoxycarbonyl)phenyl)boronic acid, 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid

A450270-1G A450270-250MG
76789-348EA 95.49 USD
76789-348 76789-350
2-Chloro-(5-methoxycarbonyl)phenylboronic acid 96%
2-Chloro-(5-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BClO₄
MW: 214.41 g/mol
MDL Number: MFCD08689528
CAS Number: 913835-92-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR