Tetrakis(triphenylphosphine)platinum(0) 98%

Supplier: Ambeed
Warning

Synonyms: Tetrakis(triphenylphosphine)platinum

A449569-1G A449569-5G
76789-346EA 20 USD
76789-346 77551-358
Tetrakis(triphenylphosphine)platinum(0) 98%
Tetrakis(triphenylphosphine)platinum(0)

Precaución: for Research Use Only

Formula: C₇₂H₆₀P₄Pt
MW: 1244.24
MDL Number: MFCD00010014
CAS Number: 14221-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR