4-Chloro-3-(tert-butylcarbamoyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-(tert-Butylcarbamoyl)-4-chlorophenyl)boronic acid

A449068-1G A449068-250MG A449068-5G
76789-338EA 129 USD
76789-338 76789-340 76789-342
4-Chloro-3-(tert-butylcarbamoyl)phenylboronic acid 97%
4-Chloro-3-(tert-butylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₃
MDL Number: MFCD07783864
CAS Number: 871332-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR