Z-Tle-OH (dicyclohexylammonium) salt 98%

Supplier: Ambeed
Warning

Synonyms: Z-tBu-Gly-OH·DCHA, Z-Tle-OH·DCHA, (-)-N-Benzyloxycarbonyl-L-tert-leucine dicyclohexyl ammonium salt, Dicyclohexylamine (S)-2-(((benzyloxy)carbonyl)amino)-3,3-dimethylbutanoate, Z-L-tert-Leucine (dicyclohexylammonium) salt

A446921-10G A446921-1G A446921-25G A446921-5G
76789-318EA 9 USD
76789-318 76789-320 76789-322 76789-324
Z-Tle-OH (dicyclohexylammonium) salt 98%
Z-Tle-OH (dicyclohexylammonium) salt

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄·C₁₂H₂₃N
MW: 446.632 g/mol
MDL Number: MFCD00161480
CAS Number: 62965-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR