Methyl 1-methyl-5-oxopyrrolidine-2-carboxylate 97%

Supplier: Ambeed
Warning

A443021-1G A443021-250MG
76789-282EA 43.5 USD
76789-282 76789-284 76789-286
Methyl 1-methyl-5-oxopyrrolidine-2-carboxylate 97%
Methyl 1-methyl-5-oxopyrrolidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MDL Number: MFCD12026428
CAS Number: 190783-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR