Pyrido[2,3-b]pyrazin-6(5H)-one 95%

Supplier: Ambeed
Warning

A440792-1G A440792-250MG
76789-274EA 79 USD
76789-274 76789-276
Pyrido[2,3-b]pyrazin-6(5H)-one 95%
Pyrido[2,3-b]pyrazin-6(5H)-one

Precaución: for Research Use Only

Formula: C7H5N3O
MDL Number: MFCD13193474
CAS Number: 35808-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR