trans-4-(Boc-aminomethyl)cyclohexylamine 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl ((trans-4-aminocyclohexyl)methyl)carbamate

A439088-5G A439088-250MG A439088-1G
76789-256EA 495 USD
76789-256 77594-372 77668-968
trans-4-(Boc-aminomethyl)cyclohexylamine 98%
trans-4-(Boc-aminomethyl)cyclohexylamine

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MW: 228.336 g/mol
MDL Number: MFCD06657670
CAS Number: 192323-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR