2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole 98%

Supplier: Ambeed
Danger

A437793-1G A437793-5G
76789-252EA 168.69 USD
76789-252 76789-254
2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole 98%
2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₉H₉ClN₂
MW: 180.64 g/mol
MDL Number: MFCD00457523
CAS Number: 4760-35-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR