3',5'-Dimethyl-4'-methoxyacetophenone 98%

Supplier: Ambeed
Warning

A435516-1G A435516-250MG
76789-238EA 107.16 USD
76789-238 76789-240
3',5'-Dimethyl-4'-methoxyacetophenone 98%
3',5'-Dimethyl-4'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.2277 g/mol
MDL Number: MFCD06201228
CAS Number: 60609-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR