2-(5-Methyl-2-nitrophenyl)acetic acid 96%

Supplier: Ambeed
Warning

A432558-1G A432558-250MG A432558-5G
76789-218EA 75.33 USD
76789-218 76789-220 76789-222
2-(5-Methyl-2-nitrophenyl)acetic acid 96%
2-(5-Methyl-2-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.17 g/mol
MDL Number: MFCD08276003
CAS Number: 37777-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR