Xanthosine 98%

Supplier: Ambeed
Warning

Synonyms: 3,9-Dihydro-9-β-D-ribofuranosyl-1H-purine-2,6-dione, 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purine-2,6-diol, 9-β-D-Ribofuranosylxanthine

A428210-100MG A428210-250MG
76789-200EA 24.41 USD
76789-200 76789-202
Xanthosine 98%
Xanthosine

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄O₆
MW: 284.23 g/mol
MDL Number: MFCD00005726
CAS Number: 146-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR