Methyl 4-bromo-2-(trifluoromethoxy)benzoate 95%

Supplier: Ambeed
Warning

A428125-1G A428125-250MG A428125-25G
76789-194EA 15.92 USD
76789-194 76789-196 76789-198
Methyl 4-bromo-2-(trifluoromethoxy)benzoate 95%
Methyl 4-bromo-2-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₃
MW: 299.041 g/mol
MDL Number: MFCD12761592
CAS Number: 933785-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR