2-Fluoro-6-(trifluoromethyl)benzophenone 95%

Supplier: Ambeed
Warning

A427606-1G A427606-250MG
76789-184EA 60.48 USD
76789-184 76789-186
2-Fluoro-6-(trifluoromethyl)benzophenone 95%
2-Fluoro-6-(trifluoromethyl)benzophenone

Precaución: for Research Use Only

Formula: C₁₄H₈F₄O
MW: 268.2063 g/mol
MDL Number: MFCD00061255
CAS Number: 208173-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR