5-Bromo-1H-1,2,4-triazole-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A427541-100MG
A427541-1G
A427541-5G
76789-182EA
29.71
USD
76789-182
77601-834
77668-906
5-Bromo-1H-1,2,4-triazole-3-carboxylic acid 98%
5-Bromo-1H-1,2,4-triazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₃H₂BrN₃O₂ MW: 191.97 g/mol |
MDL Number:
MFCD11040182 CAS Number: 674287-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...