(S)-Morpholin-2-ylmethanol 95%
Supplier: Ambeed
|
Warning
|
A425284-100MG
A425284-250MG
76789-178EA
25.47
USD
76789-178
76789-180
(S)-Morpholin-2-ylmethanol 95%
(S)-Morpholin-2-ylmethanol
Precaución: for Research Use Only
Formula:
C₅H₁₁NO₂ MW: 117.14 g/mol |
MDL Number:
MFCD09260718 CAS Number: 132073-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...