4'-Fluoro-2'-methylacetophenone 98%

Supplier: Ambeed
Warning

A424380-100G A424380-1G A424380-25G A424380-5G
76789-170EA 255 USD
76789-170 76789-172 76789-174 76789-176
4'-Fluoro-2'-methylacetophenone 98%
4'-Fluoro-2'-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₉FO
MDL Number: MFCD00028041
CAS Number: 446-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR