Z-Gly-Sar-OH 98%

Supplier: Ambeed
Warning

A423806-100MG A423806-1G A423806-250MG
76789-154EA 16 USD
76789-154 76789-156 76789-158
Z-Gly-Sar-OH 98%
Z-Gly-Sar-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₅
MW: 280.28 g/mol
MDL Number: MFCD00191104
CAS Number: 7801-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR