3-Carboxy-4-chlorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Borono-2-chlorobenzoic acid

A420086-100G A420086-10G A420086-1G A420086-250MG A420086-25G A420086-5G A420086-100MG
76789-124EA 652.46 USD
76789-124 76789-126 76789-128 76789-130 76789-132 76789-134 77668-882
3-Carboxy-4-chlorophenylboronic acid 98%
3-Carboxy-4-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₄
MW: 200.3800 g/mol
MDL Number: MFCD06801689
CAS Number: 913835-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR