3,3-Diphenylpropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,3-Diphenylpropanoic acid

A418829-100G A418829-10G A418829-25G A418829-500G A418829-5G
76789-114EA 46 USD
76789-114 76789-116 76789-118 76789-120 76789-122
3,3-Diphenylpropionic acid 98%
3,3-Diphenylpropionic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
MDL Number: MFCD00002717
CAS Number: 606-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR