2-(3-(4-Bromo-2-fluorobenzyl)-4-oxo-3,4-dihydrophthalazin-1-yl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ponalrestat
A417818-10MG
A417818-1MG
76789-110EA
41.38
USD
76789-110
76789-112
2-(3-(4-Bromo-2-fluorobenzyl)-4-oxo-3,4-dihydrophthalazin-1-yl)acetic acid 97%
2-(3-(4-Bromo-2-fluorobenzyl)-4-oxo-3,4-dihydrophthalazin-1-yl)acetic acid
Precaución: for Research Use Only
Formula:
C₁₇H₁₂BrFN₂O₃ MW: 391.19 g/mol |
MDL Number:
MFCD00204117 CAS Number: 72702-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...