Toluidine blue O Dye content, 73%+ (dye content)
Supplier: Ambeed
|
Warning
|
Synonyms:
TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride
A416842-100G
A416842-25G
A416842-5G
76789-082EA
196
USD
76789-082
76789-084
76789-086
Toluidine blue O Dye content, 73%+ (dye content)
Toluidine blue O
Precaución: for Research Use Only
Formula:
C₁₅H₁₆ClN₃S MW: 305.83 g/mol |
MDL Number:
MFCD00011934 CAS Number: 92-31-9 Merck Index: 12,09658 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...