4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-(isopropylcarbamoyl)benzeneboronic acid, 4-Chloro-3-(isopropylcarbamoyl)phenylboronic acid, (4-Chloro-3-(isopropylcarbamoyl)phenyl)boronic acid

A415542-1G A415542-250MG A415542-5G
76789-070EA 82.76 USD
76789-070 76789-072 76789-074
4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid 98%
4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BClNO₃
MW: 241.48 g/mol
MDL Number: MFCD07783866
CAS Number: 871332-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR