trans-1,2-Cyclobutanedicarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A414540-100G
A414540-1G
A414540-250MG
A414540-25G
A414540-5G
76789-060EA
740.51
USD
76789-060
76789-062
76789-064
76789-066
76789-068
trans-1,2-Cyclobutanedicarboxylic acid 97%
trans-1,2-Cyclobutanedicarboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₈O₄ MW: 144.13 g/mol |
MDL Number:
MFCD00068260 CAS Number: 1124-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...