(R)-tert-Butyldimethyl(oxiran-2-ylmethoxy)silane 97%

Supplier: Ambeed
Warning

A414484-100G A414484-1G A414484-25G A414484-5G
76789-052EA 251.44 USD
76789-052 76789-054 76789-056 76789-058
(R)-tert-Butyldimethyl(oxiran-2-ylmethoxy)silane 97%
(R)-tert-Butyldimethyl(oxiran-2-ylmethoxy)silane

Precaución: for Research Use Only

Formula: C₉H₂₀O₂Si
MW: 188.33 g/mol
MDL Number: MFCD01862147
CAS Number: 124150-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR