Triisopropyl phosphate 98%

Supplier: Ambeed
Danger

Synonyms: Triisopropylphosphate

A413016-25G A413016-1G
76789-046EA 101.85 USD
76789-046 77601-802
Triisopropyl phosphate 98%
Triisopropyl phosphate

Precaución: for Research Use Only

Formula: C₉H₂₁O₄P
MW: 224.24 g/mol
MDL Number: MFCD00015490
CAS Number: 513-02-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR