4,4'-Dimethoxy-1,1'-biphenyl 95%

Supplier: Ambeed
Warning

A412494-100G A412494-25G A412494-5G
76789-040EA 120.95 USD
76789-040 76789-042 76789-044
4,4'-Dimethoxy-1,1'-biphenyl 95%
4,4'-Dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.25 g/mol
MDL Number: MFCD00008402
CAS Number: 2132-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR