5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A411380-1G A411380-250MG A411380-5G
76789-028EA 90.18 USD
76789-028 76789-030 76789-032
5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid 95%
5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂
MW: 204.0213 g/mol
MDL Number: MFCD16659627
CAS Number: 865186-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR