Tris(2-ethylhexyl)phosphate 98%

Supplier: Ambeed
Warning

A411328-1000G A411328-100G A411328-25G A411328-500G
76789-020EA 59 USD
76789-020 76789-022 76789-024 76789-026
Tris(2-ethylhexyl)phosphate 98%
Tris(2-ethylhexyl)phosphate

Precaución: for Research Use Only

Formula: C₂₄H₅₁O₄P
MDL Number: MFCD00009491
CAS Number: 78-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR