4-Bromo-2-(methylsulfonyl)pyrimidine 98%

Supplier: Ambeed
Warning

A409027-100MG A409027-250MG A409027-1G
76789-016EA 29.71 USD
76789-016 76789-018 77594-242
4-Bromo-2-(methylsulfonyl)pyrimidine 98%
4-Bromo-2-(methylsulfonyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂S
MW: 237.0744 g/mol
MDL Number: MFCD12406118
CAS Number: 1208538-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR