3-Fluoro-5-(methylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: (3-Fluoro-5-(methylcarbamoyl)phenyl)boronic acid, 3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid

A408417-1G A408417-250MG A408417-5G
76789-006EA 81 USD
76789-006 76789-008 77668-856
3-Fluoro-5-(methylcarbamoyl)phenylboronic acid 98%
3-Fluoro-5-(methylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BFNO₃
MDL Number: MFCD07363793
CAS Number: 871332-63-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR