tert-Butyl(4-iodobutoxy)dimethylsilane 95%

Supplier: Ambeed
Warning

A404597-1G A404597-250MG A404597-5G A404597-100G A404597-25G
76788-972EA 45.62 USD
76788-972 76788-974 76788-976 77668-846 77668-848
tert-Butyl(4-iodobutoxy)dimethylsilane 95%
tert-Butyl(4-iodobutoxy)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₂₃IOSi
MW: 314.279 g/mol
MDL Number: MFCD01863695
CAS Number: 92511-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR