1,2,4-Triazole-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-1,2,4-Triazole-3-carboxylic acid
A404126-100G
A404126-1G
A404126-250MG
A404126-25G
A404126-5G
76788-960EA
328.88
USD
76788-960
76788-962
76788-964
76788-966
76788-968
1,2,4-Triazole-3-carboxylic acid 95%
1,2,4-Triazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₃H₃N₃O₂ MW: 113.0748 g/mol |
MDL Number:
MFCD00078268 CAS Number: 4928-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...