Vildagliptin 98%

Supplier: Ambeed
Warning

Synonyms: (2S)-1-((3-Hydroxyadamantan-1-yl)glycyl)pyrrolidine-2-carbonitrile

A403832-100G A403832-100MG A403832-10G A403832-1G A403832-25G A403832-5G A403832-50MG A403832-500G
76788-948EA 311.91 USD
76788-948 76788-950 76788-952 76788-954 76788-956 76788-958 77601-772 77668-840
Vildagliptin 98%
Vildagliptin

Precaución: for Research Use Only

Formula: C₁₇H₂₅N₃O₂
MW: 303.3993 g/mol
MDL Number: MFCD08275142
CAS Number: 274901-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR