Thiazolidine-2,4-dicarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Tidiacic, Tetrahydrothiazole-2,4-dicarboxylic acid

A403243-1G A403243-5G
76788-944EA 13.8 USD
76788-944 76788-946
Thiazolidine-2,4-dicarboxylic acid 98%
Thiazolidine-2,4-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₇NO₄S
MW: 177.1784 g/mol
MDL Number: MFCD00145399
CAS Number: 30097-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR