Methyl-2-amino-5-methylthiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-5-methylthiophene-3-carboxylate

A395900-100G A395900-1G A395900-5G
76788-890EA 272.66 USD
76788-890 76788-892 76788-894
Methyl-2-amino-5-methylthiophene-3-carboxylate 98%
Methyl-2-amino-5-methylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
MDL Number: MFCD01922101
CAS Number: 19369-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR