Xylene cyanole FF 75%

Supplier: Ambeed
Warning

A394410-100G A394410-10G A394410-1G A394410-25G A394410-5G
76788-880EA 305 USD
76788-880 76788-882 76788-884 76788-886 76788-888
Xylene cyanole FF 75%
Xylene cyanole FF

Precaución: for Research Use Only

Formula: C₂₅H₂₇N₂NaO₆S₂
MDL Number: MFCD00040657
CAS Number: 2650-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR