5-Bromopyridine-3,4-diamine 95%

Supplier: Ambeed
Warning

A393690-250MG A393690-25G A393690-5G
76788-874EA 27.59 USD
76788-874 76788-876 76788-878
5-Bromopyridine-3,4-diamine 95%
5-Bromopyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MW: 188.03 g/mol
MDL Number: MFCD01692479
CAS Number: 4635-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR