Pyrimidine-4-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Cyanopyrimidine

A393065-100MG A393065-1G A393065-250MG A393065-5G
76788-866EA 7 USD
76788-866 76788-868 76788-870 76788-872
Pyrimidine-4-carbonitrile 98%
Pyrimidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C5H3N3
MW: 105.0974 g/mol
MDL Number: MFCD07787613
CAS Number: 42839-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR