2,5-Dioxopyrrolidin-1-yl 2-cyanoacetate 97%

Supplier: Ambeed
Warning

A391563-100G A391563-1G A391563-25G A391563-5G
76788-864EA 514.54 USD
76788-864 77668-804 77668-806 77668-808
2,5-Dioxopyrrolidin-1-yl 2-cyanoacetate 97%
2,5-Dioxopyrrolidin-1-yl 2-cyanoacetate

Precaución: for Research Use Only

Formula: C₇H₆N₂O₄
MW: 182.13 g/mol
MDL Number: MFCD00798802
CAS Number: 56657-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR