Tributyl(tetradecyl)phosphonium chloride 98%

Supplier: Ambeed
Warning

A391219-100G A391219-25G A391219-500G A391219-5G
76788-854EA 74 USD
76788-854 76788-856 76788-858 76788-860
Tributyl(tetradecyl)phosphonium chloride 98%
Tributyl(tetradecyl)phosphonium chloride

Precaución: for Research Use Only

Formula: C₂₆H₅₆ClP
MDL Number: MFCD02683027
CAS Number: 81741-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR