2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate hydrochloride

A391201-1G A391201-250MG A391201-5G
76788-848EA 162 USD
76788-848 76788-850 76788-852
2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride 98%
2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₄·HCl
MW: 313.59 g/mol
MDL Number: MFCD06659918
CAS Number: 850567-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR