(9H-Fluoren-9-yl)methyl (trans-4-aminocyclohexyl)carbamate hydrochloride 97%

Supplier: Ambeed
Warning

A389945-100MG A389945-1G A389945-250MG
76788-840EA 37 USD
76788-840 76788-842 76788-844
(9H-Fluoren-9-yl)methyl (trans-4-aminocyclohexyl)carbamate hydrochloride 97%
(9H-Fluoren-9-yl)methyl (trans-4-aminocyclohexyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClN₂O₂
MW: 372.88 g/mol
MDL Number: MFCD03001746
CAS Number: 672310-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR