6-Bromochromen-2-one 95%

Supplier: Ambeed
Warning

A389728-10G A389728-250MG A389728-25G A389728-5G A389728-1G
76788-832EA 117 USD
76788-832 76788-834 76788-836 76788-838 77601-712
6-Bromochromen-2-one 95%
6-Bromochromen-2-one

Precaución: for Research Use Only

Formula: C₉H₅BrO₂
MDL Number: MFCD01327473
CAS Number: 19063-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR